missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Jump In™ T-REx™ HEK 293 Kit

Product Code. 13462889
Change view
Click to view available options
Quantity:
1 Kit(s), 1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13462889 1 Kit(s), 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13462889 Supplier Thermo Scientific™ Supplier No. A15008
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Jump-In™ T-REx™ HEK293 Kit allows the targeted integration of genetic material into a specific pre-engineered R4 site in the Jump-In™ HEK293 cell line, to create an inducible isogenic stable cell line with less effort and in less time than traditional cell engineering methods.

The Jump-In™ T-REx™ HEK293 Kit allows the targeted integration of genetic material into a specific pre-engineered R4 site in the Jump-In™ HEK293 cell line, to create an inducible isogenic stable cell line with less effort and in less time than traditional cell engineering methods. These cells stably express the tetracycline repressor protein, allowing for inducible expression of your retargeted gene of interest upon addition of doxycycline.

The high retargeting efficiency, made possible by the R4 sites in the HEK293 cell line, allows for the use of isogenic pools for additional experiments without the need for clonal selection. Alternatively, the high retargeting efficiency allows for the easy selection of a positive stable clone expressing your gene of interest.

The Jump-In™ T-REx™ HEK293 Kit allows users to:

  • Quickly and efficiently develop stably engineered isogenic cell pools in about half the time compared to traditional cell engineering methods
  • Inducibly express your gene of interest using doxycycline
  • Utilize isogenic expression from a defined genomic locus as the ideal solution for comparative analysis of gene families, isoforms, or orthologs
  • Generate multiple cell lines in parallel using the simplified work flow
  • Easily access the technology without complicated licenses or restrictions to interpret

Save Time with Rapid and Efficient Generation of Engineered Cell Lines
With the Jump-In™ T-REx™ HEK293 Kit you can generate functional cell pools in as little as 2 weeks without laborious clone isolation and analysis, and the streamlined protocol makes it easier to generate several cell lines at the same time. Even generation of clonal cell lines can be done with reduced time and effort due to the high percentage of positive clones. In addition, the Jump-In™ technology gives you the freedom to generate an unlimited number of cell lines without restrictive licensing requirements.

Expand Your Experimental Capabilities

  • The Jump-In™ T-REx™ HEK293 Kit allows for inducible expression of your toxic or unstable gene of interest and is the ideal solution for targets and assays where transient engineering technologies are problematic. The kit also provides a convenient way to create target panels of gene families, isoforms, or orthologs. Genes coding for large proteins or multi-unit proteins are not a problem since the Gateway™ destination vectors accept large inserts.

The kit includes:

  • Jump-In™ T-REx™ HEK293 cells, 2 x 1 mL vials
  • pJTI™ R4 Dest CMV-TO pA vector, 100 μg—A Gateway™ Destination vector that drives inducible expression of your gene of interest under control of the strong CMV promoter by inclusion of the Tet-Operon.
  • pJTI™ R4 Int vector, 100 μg—Expresses the R4 integrase to facilitate the retargeting of your gene of interest into the R4 site.

Sold separately:

  • pJTI™ R4 CMV-TO MCS pA vector (Cat. No. A15004)—A multi-site cloning vector that drives inducible expression of your gene of interest under control of the strong CMV promoter by inclusion of the Tet-Operon.
  • pJTI™ R4 EXP CMV-TO EmGFP pA vector (Cat. No. A15005)—A control vector with inducible expression of GFP under control of the strong CMV promoter by inclusion of the Tet-Operon.
TRUSTED_SUSTAINABILITY

Specifications

Description Jump In™ T-REx™ HEK 293 Kit, 1 Kit, Transfection Delivery Method, Mammalian Expression, Tetracycline Inducing Agent, 293 (HEK) Cell Line, CMV/TO Promoter, Blasticidin, Geneticin™ (G-418), Hygromycin Selection Agent, Used for Protein Expression
For Use With (Application) Protein biology, protein expression, mammalian expression, regulated expression, targeted integration, stable cell line development, Protein expression, Assay development
Cell Line 293 (HEK)
Includes Jump-In™ T-REx™ HEK293 cells (2 vials 1mL each), pJTI™ R4 DEST CMV TO pA vector (100μg), pJTI™ R4 Int vector (100μg)
Content And Storage • Jump-In™ T-REx™ HEK293 Cells, 2 x 1 mL vials (store in liquid nitrogen)
• pJTI™ R4 DEST CMV TO pA vector, 100 μg (store at -20°C)
• pJTI™ R4 Int vector, 100 μg (store at -20°C)
Delivery Type Transfection
Product Type Jump In™ T-REx™ HEK 293 Kit, HEK293 Kit
Quantity 1 Kit(s), 1 Kit
Format Kit
Promoter CMV/TO
Vector pJTI™ R4 DEST CMV TO pA , pJTI™ R4 Int
Expression System Mammalian
Inducing Agent Tetracycline
Key Functions Stable Cell Line Development, Targeted Integration, Regulated Expression
Product Line Jump-In
Selection Agent (Eukaryotic) Geneticin™ (G-418), Hygromycin, Blasticidin
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.