missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Jump In™ T-REx™ CHO-K1 Kit

Product Code. 13463259
Change view
Click to view available options
Quantity:
1 Kit(s), 1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13463259 1 Kit(s), 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13463259 Supplier Thermo Scientific™ Supplier No. A15007
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Jump-In™ T-REx™ CHO-K1 Kit allows the targeted integration of genetic material into a specific pre-engineered R4 site in the Jump-In™ CHO-K1 cell line, to create an inducible isogenic stable cell line with less effort and in less time than traditional cell engineering methods.

The Jump-In™ T-REx™ CHO-K1 Kit allows the targeted integration of genetic material into a specific pre-engineered R4 site in the Jump-In™ CHO-K1 cell line, to create an inducible isogenic stable cell line with less effort and in less time than traditional cell engineering methods. These cells stably express the tetracycline repressor protein, allowing for inducible expression of your retargeted gene of interest upon the addition of doxycycline.

The high retargeting efficiency, made possible by the R4 sites in the CHO-K1 cell line, allows the use of the isogenic pool for additional experiments without the need for clonal selection. Alternatively, the high retargeting efficiency allows for the easy selection of a positive stable clone expressing your gene of interest.

The Jump-In™ T-REx™ CHO-K1 Kit allows users to:

  • Quickly and efficiently develop stably engineered isogenic cell pools in about half the time compared to traditional cell engineering methods
  • Inducibly express your gene of interest using doxycycline
  • Utilize isogenic expression from a defined genomic locus as the ideal solution for comparative analysis of gene families, isoforms, or orthologs
  • Generate multiple cell lines in parallel using the simplified work flow
  • Easily access the technology without complicated licenses or restrictions to interpret

Save Time with Rapid and Efficient Generation of Engineered Cell Lines
With the Jump-In™ T-REx™ CHO-K1 Kit you can generate functional cell pools in as little as 2 weeks without laborious clone isolation and analysis, and the streamlined workflow makes it easier to generate several cell lines at the same time. Even generation of clonal cell lines can be done with reduced time and effort due to the high percentage of positive clones. In addition, the Jump-In™ technology gives you the freedom to generate an unlimited number of cell lines without restrictive licensing requirements.

Expand Your Experimental Capabilities
The Jump-In™ T-REx™ CHO-K1 Kit allows for inducible expression of your toxic or unstable gene of interest and is the ideal solution for targets and assays where transient engineering technologies are problematic. The kit also provides a convenient way to create target panels of gene families, isoforms, or orthologs. Genes coding for large proteins or multi-unit proteins are not a problem since the Gateway™ destination vectors accept large inserts.

The kit includes:

  • Jump-In™ T-REx™ CHO-K1 Cells, 2 x 1 mL vials
  • pJTI™ R4 Dest CMV TO pA Vector, 100 μg—A Gateway™ Destination vector that drives inducible expression of your gene of interest under control of the strong CMV promoter by inclusion of the Tet-Operon.
  • pJTI™ R4 Int Vector, 100 μg—Expresses the R4 integrase to facilitate the retargeting of your gene of interest into the R4 site.

Sold separately:

  • pJTI™ R4 CMV-TO MCS pA Vector (Cat. No. A15004)—A multi-site cloning vector that drives inducible expression of your gene of interest under control of the strong CMV promoter by inclusion of the Tet-Operon.
  • pJTI™ R4 EXP CMV-TO EmGFP pA Vector (Cat. No. A15005)—A control vector with inducible expression of GFP under control of the strong CMV promoter by inclusion of the Tet-Operon.
TRUSTED_SUSTAINABILITY

Specifications

Description Jump In™ T-REx™ CHO-K1 Kit, 1 Kit, Transfection Delivery Method, Mammalian Expression, Tetracycline Inducing Agent, CHO-K1 Cell Line, CMV/TO Promoter, Blasticidin, Geneticin™ (G-418), Hygromycin Selection Agent, Used for Protein Expression
For Use With (Application) Protein biology, protein expression, mammalian expression, regulated expression, targeted integration, stable cell line development, Assay development
Cell Line CHO-K1
Includes Jump-In™ T-REx™ CHO-K1 Cells (2 vials 1ml each), pJTI™ R4 DEST CMV TO pA vector (100μg), pJTI™ R4 Int vector (100μg)
Content And Storage • Jump-In™ T-REx™ CHO-K1 Cells, 2 x 1 mL vials (store in liquid nitrogen)
• pJTI™ R4 DEST CMV TO pA vector, 100 μg (store at -20°C)
• pJTI™ R4 Int vector, 100 μg (store at -20°C)
Delivery Type Transfection
Product Type Jump In™ T-REx™ CHO-K1 Kit, CHO-K1 Kit
Quantity 1 Kit(s), 1 Kit
Format Kit
Promoter CMV/TO
Vector pJTI™ R4 DEST CMV TO pA , pJTI™ R4 Int
Expression System Mammalian
Inducing Agent Tetracycline
Key Functions Stable Cell Line Development, Targeted Integration, Regulated Expression
Product Line Jump-In
Selection Agent (Eukaryotic) Geneticin™ (G-418), Hygromycin, Blasticidin
Show More Show Less
Why is sequential transfection recommended over co-transfection in the T-REx and GeneSwitch systems?

When a co-transfection is performed, there is no way of testing the double stable cell line for functional TetR or GeneSwitch protein, respectively. On the other hand, when sequential transfection is performed, one can functionally test the generated T-REx or GeneSwitch cell line by transiently transfecting the lacZ expression control plasmid and then picking a clone that shows the lowest basal level of expression of lacZ in the absence of the inducer, and the highest level of lacZ in the presence of the inducer. This clone can then be expanded and used to transfect the T-REx or GeneSwitch expression construct, as the case may be.

What is the main advantage of the GeneSwitch system over the T-REx system? And what is its main disadvantage?

With the GeneSwitch system, it is possible to have the absolute lowest basal levels of expression of the gene of interest, whereas the T-REx system may be a little leaky due to the inevitable presence of tetracycline in FBS. The induced level of expression in the GeneSwitch system can be even higher than that seen with the CMV promoter. The disadvantage of the GeneSwitch system is that the expression does not appear to switch off very easily in culture, although it has been demonstrated to function beautifully in transgenics. The T-REx system, on the other hand, can be switched on and off by the addition and removal of the inducer.

What is the advantage of the Flp-In T-REx system over the T-REx system?

The Flp-In T-REx system combines the targeted integration offered by the Flp-In system with the powerful inducible expression offered by the T-REx system. It allows generation of isogenic, inducible, stable cell lines and permits polyclonal selection of these cell lines. Once the Flp-In T-REx host cell line containing an integrated FRT site has been created, subsequent generation of Flp-In T-REx cell lines expressing the gene(s) of interest is rapid and efficient.

Can I use doxycycline instead of tetracycline as an inducer in the T-REx system?

Doxycycline may be used as an alternative inducing agent in the T-REx system. It is similar to tetracycline in its mechanism of action, and exhibits similar dose-response and induction characteristics as tetracycline in the T-REx system. Doxycycline has been shown to have a longer half-life than tetracycline (48 hours vs. 24 hours, respectively). We do not offer doxycycline, but it may be obtained from Sigma (Cat. No. D9891).

I am interested in a mammalian expression system where I can have regulated expression of my gene of interest. I see that you offer multiple systems for this purpose. Can you describe the main features of each system?

We offer three unique mammalian expression systems for inducible/regulated expression of the gene of interest:

- T-REx system
- Flp-In T-REx system
- GeneSwitch system

Please see below to see how they compare with one another:
System -- Basal Expression Level -- Induced Expression Level -- Response time to Maximal Expression -- Transgenic Appliation
T-Rex system -- Low -- Highest -- High -- Suitable
Flp-In T-REx system -- Lower -- High -- 24-48 hrs -- Suitable
GeneSwitch system -- Lowest -- High -- 24-48 hrs -- Suitable

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.