missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Jump-In™ GripTite™ HEK 293 Kit

Product Code. 11642089
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11642089 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11642089 Supplier Thermo Scientific™ Supplier No. A14150
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Jump-In™ GripTite™ HEK293 Kit allows the targeted integration of genetic material into a specific pre-engineered R4 site in the Jump-In™ GripTite™ HEK293 cell line, to create an isogenic stable cell line with less effort and in less time than traditional cell engineering methods.

The high retargeting efficiency, made possible by the R4 sites in the GripTite™ HEK293 cell line, allows the use of the isogenic pool for additional experiments without the need for clonal selection. Alternatively, the high retargeting efficiency allows for the easy selection of a positive stable clone expressing your gene of interest.

The Jump-In™ GripTite™ HEK293 Kit Lets You:
• Quickly and efficiently develop stably engineered isogenic cell pools in about half the time compared to traditional cell engineering methods
• Utilize isogenic expression from a defined genomic locus as the ideal solution for comparative analysis of gene families, isoforms, or orthologs
• Generate multiple cell lines in parallel using the simplified work flow
• Easily access the technology without complicated licenses or restrictions to interpret

Save Time with Rapid and Efficient Generation of Engineered Cell Lines
With the Jump-In™ GripTite™ HEK293 Kit you can generate functional cell pools in as little as 2 weeks without laborious clone isolation and analysis, and the streamlined protocol makes it easier to generate several cell lines at the same time. Even generation of clonal cell lines can be done with reduced time and effort due to the high percentage of positive clones. In addition, the Jump-In™ technology gives you the freedom to generate an unlimited number of cell lines without restrictive licensing requirements.

Expand Your Experimental Capabilities
The Jump-In™GripTite™ HEK293 Kit is the ideal solution for cells and assays where transient engineering technologies are problematic, as well as for difficult to 'engineer' cell lines. The kit also provides a convenient way to create target panels of gene families, isoforms, or orthologs. Genes coding for large proteins or multi-unit proteins are not a problem since the Gateway™ destination vectors accept large inserts.

The kit includes:
• pJTI™ R4 Dest CMV pA vector (100 μg)
• pJTI™ R4 Int vector (100 μg)
• Jump-In™GripTite™ HEK293 Cells (2 vials @ 1 ml each)

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line Jump-In™ GripTite™ Hek293
Content And Storage The kit includes:
Jump-In™ GripTite™ HEK293 cells
pJTI™ R4 Int (integrase vector)
pJTI™ R4 DEST CMV pA (destination vector)

Upon receipt, immediately store the cells in liquid nitrogen and the vectors at -20°C. Cells stored at -80°C can quickly lose viability.
Delivery Type Transfection
Format Kit
Promoter CMV
Expression System Mammalian
For Use With (Application) Protein expression, Assay development
Key Functions Stable Cell Line Development, Targeted Integration
Product Line Jump-In
Product Type HEK293 Kit
Quantity 1 Kit
Selection Agent (Eukaryotic) Geneticin™ (G-418), Hygromycin
System Type Jump-In™
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.