missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Jump-In™ Fast Gateway™ System

Product Code. 10358814
Click to view available options
Quantity:
20 Reactions
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10358814

Brand: Invitrogen™ A10893

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Jump-In™ Fast Gateway™ System

The Jump-In™ Fast Gateway™ System is intended for rapid engineering of mammalian cell lines through the stable and irreversible integration of your gene of interest (GOI) at specific genomic loci (pseudo attP sites). Following stable integration, only 5-10 clones need to be analyzed to identify clones with high expression levels.
The Jump-In™ Fast Gateway™ System consists of:

  • The Jump-In™ Fast Gateway™ Core Kit containing the promoter-less pJTI™ Fast DEST expression vector and a vector for transient expression of the PhiC31 integrase.
  • The MultiSite Gateway™ Pro Plus kit for cloning of up to 4 gene fragments into the promoter-less pJTI™ Fast DEST expression vector.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Constitutive or Inducible System Constitutive
    Format Liquid
    Description Jump-In™ Fast Gateway™ System, 20 rxns, Liquid, Untagged Protein Tag, Transfection Delivery Method, Jump-In Vectors, pJTI Vector Type, Gateway™ Cloning Method, Used in Stem Protein Expression, Mammalian Expression, HygroR Selection Marker (Eukaryotic)
    Promoter None (Promoterless)
    Product Type Jump-In™ Fast Gateway™ System
    Selection Agent (Eukaryotic) Hygromycin
    Content And Storage -20°C
    Protein Tag Untagged
    Quantity 20 Reactions
    Vector pJTI, Jump-in vectors
    For Use With (Application) Protein biology, protein expression, mammalian expression, targeted integration, stable cell line development
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.