missing translation for 'onlineSavingsMsg'
Learn More

Jumonji/JARID2 Antibody [DyLight 350], Novus Biologicals™

Product Code. 30735025 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735025 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735025 Supplier Novus Biologicals Supplier No. AF6090D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Jumonji/JARID2 Polyclonal antibody specifically detects Jumonji/JARID2 in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Jumonji/JARID2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias JMJjumonji-like protein, jumonji (mouse) homolog, jumonji homolog, jumonji, AT rich interactive domain 2, Jumonji/ARID domain-containing protein 2, protein Jumonji
Host Species Sheep
Immunogen E. coli-derived recombinant human Jumonji/JARID2, Met1-Thr159, Accession # Q92833
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Epigenetics, Neuroscience, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3720
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.