missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jumonji/JARID2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 17 publications
£183.00 - £379.00
Specifications
| Antigen | Jumonji/JARID2 |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1:500, Simple Western 1:10, Chromatin Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 22396653), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:1000, Immunoprecipitation 1:10-1:500. Use reported in scientific literature, Immunohistochemistry-Paraffin 1:200, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockout Validated |
| Applications | Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18006495
|
Novus Biologicals
NB100-2214SS |
0.025 mL |
£183.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18741134
|
Novus Biologicals
NB100-2214 |
0.1 mL | |||||||
Description
Jumonji/JARID2 Polyclonal specifically detects Jumonji/JARID2 in Human, Mouse, Rabbit samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
| Jumonji/JARID2 | |
| Western Blot 1:500, Simple Western 1:10, Chromatin Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 22396653), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:1000, Immunoprecipitation 1:10-1:500. Use reported in scientific literature, Immunohistochemistry-Paraffin 1:200, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockout Validated | |
| Polyclonal | |
| Rabbit | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Neuroscience, Transcription Factors and Regulators | |
| Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide | |
| JMJjumonji-like protein, jumonji (mouse) homolog, jumonji homolog, jumonji, AT rich interactive domain 2, Jumonji/ARID domain-containing protein 2, protein Jumonji | |
| JARID2 | |
| IgG | |
| Affinity Purified | |
| Human and mouse. Rabbit reactivity reported in scientific literature (PMID: 24074864). |
| 1.0 mg/ml | |
| Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rabbit | |
| Q92833 | |
| 3720 | |
| A synthetic peptide made to an N-terminal portion of the human JARID2 protein sequence (between residues 1-100). [UniProt# Q92833] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title