missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Jumbosep™ Centrifugal Device Membrane Inserts
Click to view available options
MWCO:
10 kDa
100 kDa
3 kDa
30 kDa
300 kDa
Color:
Blue
Clear
Gray
Orange
Red
Unit Size:
Pack of 12
Description
- Typically concentrates 60 mL sample volumes to 5 mL in 30 minutes
- Achieve 50X concentration and > 90% recovery in just minutes
- Low protein-binding Omega™ membrane and polysulfone housing minimize losses due to non-specific binding
- Versatile Omega membrane is available in a variety of MWCOs
- Color-coded and laser etched for easy identification
- Built-in dead stop prevents spinning to dryness
- Unique sealing mechanism prevents retentate leakage and filtrate contamination
- Economical - Sample reservoir and filtrate receiver can be sanitized or autoclaved, and reused
Specifications
Specifications
| Color | Orange |
| Description | EFA: 15.2 cm2 |
| Material (Membrane) | PES |
| MWCO | 300 kDa |
| Sterility | Non-sterile |
| Quantity | 12/Pk. |
| Volume (Metric) | 20 to 60 mL (Sample) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction