Learn More
Trajan™ Jumbo Syringe
Product Code. 10300811
Change view
Click to view available options
Volume (Metric):
500 mL
1000 mL
2000 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10300811 500 mL Each
10751042 1000 mL Each
10045470 2000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10300811 Supplier Trajan™ Supplier No. 009910
Only null left
Add to basket
Add to basket
Hold and dispense large gas volumes. Lightweight and ideal for medical equipment calibration.
  • Designed for holding and dispensing large volumes of gas
  • Heavy duty acrylic barrels
  • Easy access to sample for the addition of standards or removal of subsample via secondary port
  • Plunger stem can be unscrewed for ease of transportation and storage
  • Ideal for calibration of medical equipment such as respirators and spirometers, and for stack and air sampling.
Specifications
Material Acrylic, Acrylic (Barrel)
For Use With (Application) For calibration of medical equipment such as respirators and spirometers, and for stack and air sampling
Product Type Jumbo Syringe
Model 500MAR-LL-GT
Volume (Metric) 500 mL