missing translation for 'onlineSavingsMsg'
Learn More

Julolidine, 98%

Product Code. p-7026769
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11496126

Brand: Thermo Scientific Alfa Aesar B22976.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 479-59-4 | C12H15N | 173.259 g/mol

Julolidine used as amine building block and in chemoluminescent dye. Also used in photoconductive materials, chemiluminescence substances, chromogenic substrates in analytical redox reactions, dye intermediates, potential antidepressants and tranquilizers, nonlinear optical materials, high sensitivity photopolymerizable materials, and for improving color stability in photography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Julolidine used as amine building block and in chemoluminescent dye. Also used in photoconductive materials, chemiluminescence substances, chromogenic substrates in analytical redox reactions, dye intermediates, potential antidepressants and tranquilizers, nonlinear optical materials, high sensitivity photopolymerizable materials, and for improving color stability in photography.

Solubility
Soluble in toluene. Insoluble in water.

Notes
Air sensitive. Store away from air, oxidizing agents, carbon dioxide. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 479-59-4
Molecular Formula C12H15N
Molecular Weight (g/mol) 173.259
MDL Number MFCD00006917
InChI Key DZFWNZJKBJOGFQ-UHFFFAOYSA-N
Synonym julolidine, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizine, 1h,5h-benzo ij quinolizine, 2,3,6,7-tetrahydro, 1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinoline, 2,3,6,7-tetrahydro-1h,5h-benzo i,j quinolizine, 2,3,6,7-tetrahydro-1h,5h-pyrido 3,2,1-ij quinoline, 2,6,7-tetrahydro-1h,5h-benzo ij quinolizine
PubChem CID 68069
SMILES C1CC2=C3C(=CC=C2)CCCN3C1

Specifications

Melting Point 32°C to 37°C
Boiling Point 170°C to 173°C (31 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Beilstein 139925
Sensitivity Air sensitive
Solubility Information Soluble in toluene. Insoluble in water.
Formula Weight 173.26
Percent Purity 98%
Chemical Name or Material Julolidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.