missing translation for 'onlineSavingsMsg'
Learn More

Julolidine, 98%

Product Code. 11496126
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496126 1 g 1g
11406136 5 g 5g
11416136 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496126 Supplier Thermo Scientific Alfa Aesar Supplier No. B22976.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 479-59-4 | C12H15N | 173.259 g/mol

Julolidine used as amine building block and in chemoluminescent dye. Also used in photoconductive materials, chemiluminescence substances, chromogenic substrates in analytical redox reactions, dye intermediates, potential antidepressants and tranquilizers, nonlinear optical materials, high sensitivity photopolymerizable materials, and for improving color stability in photography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Julolidine used as amine building block and in chemoluminescent dye. Also used in photoconductive materials, chemiluminescence substances, chromogenic substrates in analytical redox reactions, dye intermediates, potential antidepressants and tranquilizers, nonlinear optical materials, high sensitivity photopolymerizable materials, and for improving color stability in photography.

Solubility
Soluble in toluene. Insoluble in water.

Notes
Air sensitive. Store away from air, oxidizing agents, carbon dioxide. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 479-59-4
Molecular Formula C12H15N
Molecular Weight (g/mol) 173.259
MDL Number MFCD00006917
InChI Key DZFWNZJKBJOGFQ-UHFFFAOYSA-N
Synonym julolidine, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizine, 1h,5h-benzo ij quinolizine, 2,3,6,7-tetrahydro, 1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinoline, 2,3,6,7-tetrahydro-1h,5h-benzo i,j quinolizine, 2,3,6,7-tetrahydro-1h,5h-pyrido 3,2,1-ij quinoline, 2,6,7-tetrahydro-1h,5h-benzo ij quinolizine
PubChem CID 68069
SMILES C1CC2=C3C(=CC=C2)CCCN3C1

Specifications

Melting Point 32°C to 37°C
Boiling Point 170°C to 173°C (31 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
Beilstein 139925
Sensitivity Air sensitive
Solubility Information Soluble in toluene. Insoluble in water.
Formula Weight 173.26
Percent Purity 98%
Chemical Name or Material Julolidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.