missing translation for 'onlineSavingsMsg'
Learn More

Jujuboside A, MedChemExpress

Product Code. 16493886
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493886 25 mg 25mg
16491468 5 mg 5mg
16439058 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16493886 Supplier Medchem Express Supplier No. HYN0659
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Jujuboside A is a glycoside extracted from Semen Ziziphi Spinosae, a Chinese herbal medicine used to treat insomnia and anxiety.

Jujuboside A is a glycoside extracted from Semen Ziziphi Spinosae, a Chinese herbal medicine used to treat insomnia and anxiety.

Specifications

Chemical Name or Material Jujuboside A
CAS 55466-04-1
Color White
Purity Grade Notes Research
Molecular Formula C58H94O26
Quantity 25 mg
Solubility Information DMSO : 100 mg/mL (82.83 mM; Need ultrasonic) ∣H2O : 50 mg/mL (41.41 mM; Need ultrasonic)
SMILES C[C@]([C@]1([H])CC2)(CC[C@]3([H])[C@@]1(CC[C@H](O[C@@](OC[C@H](O)[C@@H]4O[C@@](O[C@H](CO[C@@H]([C@@H]([C@@H](O)[C@@H]5O)O)O[C@@H]5CO)[C@@H](O)[C@@H]6O)([H])[C@@H]6O[C@@](OC[C@@H](O)[C@@H]7O)([H])[C@@H]7O)([H])[C@@H]4O[C@@](O[C@@H](C)[C@H](O)[C@H]8O)([H])[C@@H]8O)C3(C)C)C)[C@@]9(CO%10)[C@@]2([H])[C@]([C@]%11(O)C)([H])[C@@]%10(O[C@@H](/C=C(C)/C)C%11)C9
Molecular Weight (g/mol) 1207.35
Formula Weight 1207.35
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.