missing translation for 'onlineSavingsMsg'
Learn More

JTV 519 Fumarate, TRC

Product Code. 30400122
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400122 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400122 Supplier Toronto Research Chemicals Supplier No. TRCJ2120131MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material JTV 519 Fumarate
CAS 1883549-36-7
Molecular Formula C25H32N2O2 • (C4H4O4)
Quantity 1 mg
Synonym 1-(2,3-Dihydro-7-methoxy-1,4-benzothiazepin-4(5H)-yl)-3-[4-(phenylmethyl)-1-piperidinyl]-1-propanone, 2-Butenedioate (1:1)
InChI Formula InChI=1S/C25H32N2O2S.C4H4O4/c1-29-23-7-8-24-22(18-23)19-27(15-16-30-24)25(28)11-14-26-12-9-21(10-13-26)17-20-5-3-2-4-6-20;5-3(6)1-2-4(7)8/h2-8,18,21H,9-17,19H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1+
SMILES O=C(N1CCSC2=CC=C(C=C2C1)OC)CCN3CCC(CC3)CC4=CC=CC=C4.O=C(O)/C=C/C(O)=O
Molecular Weight (g/mol) 424.6011607
Formula Weight 540.2294
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.