missing translation for 'onlineSavingsMsg'
Learn More

JTE 013, TRC

Product Code. 30399800
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399800 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399800 Supplier Toronto Research Chemicals Supplier No. TRCJ21198325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(2,6-dichloro-4-pyridyl)-3-[(4-isopropyl-1,3-dimethyl-pyrazolo[3,4-b]pyridin-6-yl)amino]urea
CAS 383150-41-2
Molecular Formula C17H19Cl2N7O
Quantity 25 mg
Synonym JTE 013
InChI Formula InChI=1S/C17H19Cl2N7O/c1-8(2)11-7-14(22-16-15(11)9(3)25-26(16)4)23-24-17(27)20-10-5-12(18)21-13(19)6-10/h5-8H,1-4H3,(H,22,23)(H2,20,21,24,27)
SMILES CC(C)c1cc(NNC(=O)Nc2cc(Cl)nc(Cl)c2)nc3c1c(C)nn3C
IUPAC Name 1-(2,6-dichloropyridin-4-yl)-3-[(1,3-dimethyl-4-propan-2-ylpyrazolo[3,4-b]pyridin-6-yl)amino]urea
Molecular Weight (g/mol) 408.29
Formula Weight 407.1028
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.