missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JTB Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | JTB |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18374956
|
Novus Biologicals
NBP3-04953-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18367384
|
Novus Biologicals
NBP3-04953-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
JTB Polyclonal specifically detects JTB in Human, Rat samples. It is validated for Western Blot.Specifications
| JTB | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| hJT, HJTB, HSPC222, jumping translocation breakpoint, Jumping translocation breakpoint protein, PAR, PAR protein, Prostate androgen-regulated protein, protein JTB | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 31-105 of human JTB (NP_006685.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS with 50% glycerol, pH7.3. | |
| 10899 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title