missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JTB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552307
This item is not returnable.
View return policy
Description
Immunogen sequence: EEKLSASTSN LPCWLVEEFV VAEECSPCSN FRAKTTPECG PTGYVEKITC SSSKRNEFKS CRSALMEQRL Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 86%.
Required for normal cytokinesis during mitosis. Plays a role in the regulation of cell proliferation. May be a component of the chromosomal passenger complex (CPC), a complex that acts as a key regulator of mitosis. The CPC complex has essential functions at the centromere in ensuring correct chromosome alignment and segregation and is required for chromatin-induced microtubule stabilization and spindle assembly. Increases AURKB activity. Inhibits apoptosis induced by TGFB1. Overexpression induces swelling of mitochondria and reduces mitochondrial membrane potential.
Specifications
| JTB | |
| Polyclonal | |
| Unconjugated | |
| JTB | |
| hJT; HJTB; HSPC222; Jtb; jumping translocation breakpoint; jumping translocation breakpoint protein; PAR; PAR protein; Prostate androgen-regulated protein; protein JTB | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10899 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O76095 | |
| JTB | |
| Recombinant protein corresponding to Human JTB. Recombinant protein control fragment (Product #RP-89171). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction