missing translation for 'onlineSavingsMsg'
Learn More

JT010, MedChemExpress

Product Code. 16487127
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487127 5 mg 5mg
16474408 10 mg 10mg
16433066 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16487127 Supplier Medchem Express Supplier No. HY111132
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress JT010 is a potent agonist of TRPA1 with an EC50 of 0.65 nM.

JT010 is a potent agonist of TRPA1 with an EC50 of 0.65 nM.

Specifications

Chemical Name or Material JT010
CAS 917562-33-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H19ClN2O3S
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (281.81 mM; Need ultrasonic)
SMILES O=C(N(C1=NC(C2=CC=C(OC)C=C2)=CS1)CCCOC)CCl
Molecular Weight (g/mol) 354.85
Formula Weight 354.85
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.