missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ (+)-JQ1 maleimide
Description
(+)-JQ1 maleimide is a probe that comprises a cysteine-reactive maleimide connected to the BRD4 ligand, JQ1, via a PEG2 linker. It can be used in the COFFEE method (COvalent Functionalization Followed by E3 Electroporation) in which (+)-JQ1 maleimide is covalently attached to VHL followed by electroporation into live cells. The E3 ligase forms a complex with BRD4 intracellularly and induces its degradation.
Specifications
Specifications
| Chemical Name or Material | (S)-N-(2-(2-(2-(2-(4-(4-Chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)acetamido)ethoxy)ethoxy)ethyl)-6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanamide |
| Quantity | 10 mg |
| Target | Active Degraders |
| Molecular Formula | C35H42ClN7O6S |
| Purity | 95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?