missing translation for 'onlineSavingsMsg'
Learn More

Imrecoxib, MedChemExpress

Product Code. 16421067
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16421067

Brand: Medchem Express HY114200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Imrecoxib (BAP-909) is a novel and selective cyclooxygenase 2 (COX-2) inhibitor with an IC50 value of 18 nM, it also inhibits COX1- activity with an IC50 value of 115 nM. Imrecoxib (BAP-909) has anti-inflammatory effect.

Imrecoxib (BAP-909) is a novel and selective cyclooxygenase 2 (COX-2) inhibitor with an IC50 value of 18 nM, it also inhibits COX1- activity with an IC50 value of 115 nM. Imrecoxib (BAP-909) has anti-inflammatory effect.

Specifications

Chemical Name or Material Imrecoxib
CAS 395683-14-4
Color White
Purity Grade Notes Research
Molecular Formula C21H23NO3S
Quantity 50 mg
Synonym BAP-909
Solubility Information DMSO : 100 mg/mL (270.65 mM; Need ultrasonic)
SMILES O=C1N(CCC)CC(C2=CC=C(S(=O)(C)=O)C=C2)=C1C3=CC=C(C)C=C3
Molecular Weight (g/mol) 369.48
Formula Weight 369.48
Percent Purity 98.64%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.