missing translation for 'onlineSavingsMsg'
Learn More

JQ-1 (carboxylic acid), MedChemExpress

Product Code. 16491956
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16491956

Brand: Medchem Express HY78695

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress JQ-1 carboxylic acid, a type of (+)-JQ-1 derivative, is a potent BET bromodomain inhibitor for downregulating PD-L1 expression on the surface of tumor cells. JQ-1 carboxylic acid can be used as a precursor to synthesize PROTACs, which targets BET bromodomains.

JQ-1 carboxylic acid, a type of (+)-JQ-1 derivative, is a potent BET bromodomain inhibitor for downregulating PD-L1 expression on the surface of tumor cells. JQ-1 carboxylic acid can be used as a precursor to synthesize PROTACs, which targets BET bromodomains.

Specifications

Chemical Name or Material JQ-1 (carboxylic acid)
Physical Form Powder
CAS 202592-23-2
Color Yellow
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C19H17ClN4O2S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 31.25 mg/mL (77.95 mM; ultrasonic and warming and heat to 60°C)
SMILES ClC1=CC=C(C(C2=C(N3C4=NN=C3C)SC(C)=C2C)=N[C@H]4CC(O)=O)C=C1
Molecular Weight (g/mol) 400.88
Formula Weight 400.88
Percent Purity 98.01%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.