missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
JPH3 Polyclonal antibody specifically detects JPH3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Applications | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CAGL237, junctophilin 3, junctophilin type 3 |
| Host Species | Rabbit |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Target Species | Mouse, Rat |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?