missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ JPH2 Polyclonal Antibody

Product Code. 12808792
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12808792 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12808792 Supplier Invitrogen™ Supplier No. PA520641

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is 293 cell lysate. PA5-20641 can be used with blocking peptide PEP-0761.

This gene encodes a protein with a leucine-rich repeat and a calponin homology domain. Polymorphism in this gene may be associated with susceptibililty to knee osteoarthritis. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JPH2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene JPH2
Gene Accession No. Q2PS20, Q9BR39, Q9ET78
Gene Alias 1110002E14Rik; CMH17; Jp2; JP-2; JP2NT; JPH2; jph2 {ECO:0000312; junctophilin 2; junctophilin type 2; Junctophilin-2; Junctophilin-2 N-terminal fragment; RGD:1305196}
Gene Symbols JPH2
Host Species Rabbit
Immunogen A 14 amino acid peptide near the carboxy terminus of human JPH2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296345, 57158, 59091
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.