missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ JP 1302 dihydrochloride
GREENER_CHOICE

Product Code. 18085582
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18085582 50 mg 50mg
18005492 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18085582 Supplier Tocris Bioscience™ Supplier No. 2666/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective α2C antagonist

α2C-adrenoceptor antagonist that displays ∽ 50-fold selectivity over other α2-adrenoceptor subtypes (Ki values are 28, 1470, 1700 and 3150 nM for human α2C, α2B, α2D and α2A subtypes respectively). Potently antagonizes adrenalin-stimulated 35GTPγS binding in vitro (KB = 16 nM) and produces antidepressant and antipsychotic-like effects in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1259314-65-2
Molecular Formula C24H26Cl2N4

Specifications

Quantity 50 mg
Formula Weight 441.4
Purity >99%
Molecular Weight (g/mol) 441.4
Chemical Name or Material JP 1302 dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.