missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ JP 1302 dihydrochloride

Description
α2C-adrenoceptor antagonist that displays ∽ 50-fold selectivity over other α2-adrenoceptor subtypes (Ki values are 28, 1470, 1700 and 3150 nM for human α2C, α2B, α2D and α2A subtypes respectively). Potently antagonizes adrenalin-stimulated 35GTPγS binding in vitro (KB = 16 nM) and produces antidepressant and antipsychotic-like effects in vivo.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 441.4 |
| Purity | >99% |
| Molecular Weight (g/mol) | 441.4 |
| Chemical Name or Material | JP 1302 dihydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?