Learn More
Edulab™ Joulemeter/Wattmeter Light
Product Code. 12806472
Product Code. 12806472
Brand: Edulab™ 211001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
compact unit, can measure energy taken up by a load, either AC or DC power provided with pair of 4mm sockets, digital display 110-220V 50/60Hz blue 208mm x 155mm x 115mm 0V to 15V AC, 1V to 210V DC, 10A AC/DC Edu-Lab