missing translation for 'onlineSavingsMsg'
Learn More

Josamycin Propionate, TRC

Product Code. 30393364
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393364 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393364 Supplier Toronto Research Chemicals Supplier No. TRCJ21203310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material [(2S,3S,4R,6S)-6-[(2R,3S,4R,5R,6S)-6-[((4R,5S,6S,7R,9R,10R,11E,13E,16R)-4-acetyloxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)-10-propanoyloxy-1-oxacyclohexadeca-11,13-dien-6-yl)oxy]-4-(dimethylamino)-5-hydroxy-2-methyloxan-3-yl]oxy-4-hydroxy-2,4-dimeth
CAS 40922-77-8
Molecular Formula C45 H73 N O16
Quantity 10 mg
Synonym Josamycin propionate, 10-Propionyljosamycin, 9-Propionyljosamycin, Josamy, Josamycin Propionate, Josaxin
InChI Formula InChI=1S/C45H73NO16/c1-13-34(49)59-32-18-16-14-15-17-27(5)55-36(51)23-33(58-30(8)48)42(54-12)41(31(19-20-47)22-26(32)4)62-44-39(52)38(46(10)11)40(28(6)57-44)61-37-24-45(9,53)43(29(7)56-37)60-35(50)21-25(2)3/h14-16,18,20,25-29,31-33,37-44,52-53H,13,17,19,2
SMILES CCC(=O)O[C@H]1\C=C\C=C\C[C@@H](C)OC(=O)C[C@@H](OC(=O)C)[C@H](OC)[C@@H](O[C@@H]2O[C@H](C)[C@@H](O[C@H]3C[C@@](C)(O)[C@@H](OC(=O)CC(C)C)[C@H](C)O3)[C@@H]([C@H]2O)N(C)C)[C@@H](CC=O)C[C@H]1C
IUPAC Name [(2S,3S,4R,6S)-6-[(2R,3S,4R,5R,6S)-6-[[(4R,5S,6S,7R,9R,10R,11E,13E,16R)-4-acetyloxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)-10-propanoyloxy-1-oxacyclohexadeca-11,13-dien-6-yl]oxy]-4-(dimethylamino)-5-hydroxy-2-methyloxan-3-yl]oxy-4-hydroxy-2,4-dimeth
Molecular Weight (g/mol) 884.058
Formula Weight 883.493
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.