missing translation for 'onlineSavingsMsg'
Learn More

Josamycin, MedChemExpress

Product Code. 16478826
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478826 100 mg 100mg
16423816 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478826 Supplier Medchem Express Supplier No. HYB1920
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Josamycin (EN-141) is a macrolide antibiotic exhibiting antimicrobial activity against a wide spectrum of pathogens, such as bacteria. The dissociation constant Kd from ribosome for Josamycin is 5.5 nM.

Josamycin (EN-141) is a macrolide antibiotic exhibiting antimicrobial activity against a wide spectrum of pathogens, such as bacteria. The dissociation constant Kd from ribosome for Josamycin is 5.5 nM.

Specifications

Chemical Name or Material Josamycin
Physical Form Powder
CAS 16846-24-5
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C42H69NO15
For Use With (Application) COVID-19-immunoregulation
Synonym EN-141
Solubility Information DMSO : ≥ 41.67 mg/mL (50.33 mM) ∣H2O : 8.33 mg/mL (10.06 mM; ultrasonic and adjust pH to 6 with HCl)
SMILES O[C@H]1[C@](O[C@H](C)[C@@H](O[C@@](O[C@@H](C)[C@@H]2OC(CC(C)C)=O)([H])C[C@]2(O)C)[C@@H]1N(C)C)([H])O[C@@H]([C@H](C[C@H]([C@H](/C=C/C=C/C3)O)C)CC=O)[C@H]([C@](CC(O[C@@H]3C)=O)([H])OC(C)=O)OC
Molecular Weight (g/mol) 827.99
Formula Weight 827.99
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.