missing translation for 'onlineSavingsMsg'
Learn More

Josamycin, 98+%, Thermo Scientific™

 

Brand:  Thermo Scientific Alfa Aesar J62245.MF

Quantity 50mg

Product Code. 15414659

  • £123.00 / 50mg
 Restricted Item
Estimated Shipment: 21-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Josamycin, is used as a macrolide antibiotic effective against a variety of pathogens.

Solubility
Soluble in ethanol

Notes
Protect from light and moisture. Store in cool place. Keep container tightl
TRUSTED_SUSTAINABILITY
Specifications

Specifications

16846-24-5
50mg
MFCD00210320
Leucomycin A3
XJSFLOJWULLJQS-NGVXBBESSA-N
(2S,3S,4R,6S)-6-{[(2R,3S,4R,5R,6S)-6-{[(4R,5S,6S,7R,9R,10R,11E,13E,16R)-4-(acetyloxy)-10-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)-1-oxacyclohexadeca-11,13-dien-6-yl]oxy}-4-(dimethylamino)-5-hydroxy-2-methyloxan-3-yl]oxy}-4-hydroxy-2,4-dimethyloxan-3-yl 3-methylbutanoate
131849444
≥98%
White
C42H69NO15
14,5267
Soluble in ethanol
CO[C@H]1[C@@H](CC(=O)O[C@H](C)C\C=C\C=C\[C@H](O)[C@H](C)C[C@H](CC=O)[C@@H]1O[C@@H]1O[C@H](C)[C@@H](O[C@H]2C[C@@](C)(O)[C@@H](OC(=O)CC(C)C)[C@H](C)O2)[C@@H]([C@H]1O)N(C)C)OC(C)=O
828.01
827.99
Josamycin
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.