missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JNK3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599452
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total JNK3.
MAPK10 (JNK3) is a neuronal-specific form of c-Jun N-terminal kinases (JNKs). MAPK10 is involved in neuronal apoptosis and phosphorylates MAP2K4. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. This protein is a neuronal-specific form of c-Jun N-terminal kinases (JNKs). Through its phosphorylation and nuclear localization, this kinase plays regulatory roles in the signaling pathways during neuronal apoptosis. Beta-arrestin 2, a receptor-regulated MAP kinase scaffold protein, is found to interact with, and stimulate the phosphorylation of this kinase by MAP kinase kinase 4 (NhpK4). Cyclin-dependent kianse 5 can phosphorylate, and inhibit the activity of this kinase, which may be important in preventing neuronal apoptosis. Four alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
| JNK3 | |
| Polyclonal | |
| Unconjugated | |
| MAPK10 | |
| C230008H04Rik; c-Jun N-terminal kinase 3; hypothetical protein LOC569698; jnk3; JNK3 alpha protein kinase; JNK3 beta1 protein kinase; JNK3 beta2 protein kinase; JNK3A; JNK3B1; JNK3B2; MAP kinase 10; MAP kinase p49 3F12; MAPK 10; MAPK10; mitogen activated protein kinase 10; mitogen-activated protein kinase 10; OTTHUMP00000161181; OTTHUMP00000161183; OTTHUMP00000218718; p493F12; p54-beta; p54bSAPK; PRKM10; SAPb; SAPK(beta); SAPK/Erk/kinase 2; SAPK1B; SAPK-beta; SAPKC; Serk2; Stress activated protein kinase beta; stress-activated protein kinase 1b; Stress-activated protein kinase JNK3; zgc:123234 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 25272, 26414, 5602 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P49187, P53779, Q61831 | |
| MAPK10 | |
| A synthesized peptide derived from human MAPK10(Accession P53779), corresponding to amino acid residues P371-V421. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction