missing translation for 'onlineSavingsMsg'
Learn More

JNK3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£145.00 - £417.00

Specifications

Antigen JNK3
Dilution Western Blot 1:500 - 1:2000, ELISA
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30232415
View Documents
Novus Biologicals
NBP3-37911-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30227418
View Documents
Novus Biologicals
NBP3-37911-20ul
20 μL
£145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

JNK3 Polyclonal antibody specifically detects JNK3 in Mouse samples. It is validated for ELISA,Western Blot
Specifications

Specifications

JNK3
ELISA, Western Blot
Unconjugated
Rabbit
Immunology, Innate Immunity, Protein Kinase, Wnt Signaling Pathway
PBS (pH 7.3), 50% glycerol
5602
IgG
Affinity purified
Western Blot 1:500 - 1:2000, ELISA
Polyclonal
Purified
RUO
Mouse
c-Jun N-terminal kinase 3, EC 2.7.11, EC 2.7.11.24, JNK3 alpha protein kinase, JNK3A, JNK3FLJ12099, MAP kinase 10, MAP kinase p49 3F12, MAPK 10, MGC50974, mitogen-activated protein kinase 10, p493F12, p54bSAPK, PRKM10FLJ33785, stress activated protein kinase beta, Stress-activated protein kinase JNK3
Recombinant fusion protein containing a sequence corresponding to amino acids 175-464 of human JNK3 (NP_620448.1).,, Sequence:, CGIKHLHSAGIIHRDLKPSNIVVKSDCTLKILDFGLARTAGTSFMMTPYVVTRYYRAPEVILGMGYKENVDIWSVGCIMGEMVRHKILFPGRDYIDQWNKVIEQLGTPCPEFMKKLQPTVRNYVENRPKYAGLTFPKLFPDSLFPADSEHNKLKASQARDLLSKMLVIDPAKRISVDDALQHPYINVWYDPAEVEAPPPQIYDKQLDEREHTIEEWKELIYKEVMNSEEKTKNGVVKGQPSPSGAAVNSSESLPPSSSVNDISSMSTDQTLASDTDSSLEASAGPLGCCR
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.