missing translation for 'onlineSavingsMsg'
Learn More

JNK3 Antibody (4E12-1F7), Novus Biologicals™

Product Code. 18348968 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348968 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18348968 Supplier Novus Biologicals Supplier No. H00005602M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

JNK3 Monoclonal antibody specifically detects JNK3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Proximity Ligation Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen JNK3
Applications Western Blot, ELISA, Immunocytochemistry, Proximity Ligation Assay
Classification Monoclonal
Clone 4E12-1F7
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH65516
Gene Alias c-Jun N-terminal kinase 3, EC 2.7.11, EC 2.7.11.24, JNK3 alpha protein kinase, JNK3A, JNK3FLJ12099, MAP kinase 10, MAP kinase p49 3F12, MAPK 10, MGC50974, mitogen-activated protein kinase 10, p493F12, p54bSAPK, PRKM10FLJ33785, stress activated protein kinase beta, Stress-activated protein kinase JNK3
Host Species Mouse
Immunogen MAPK10 (AAH65516.1, 1 a.a. ~ 319 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MELMDANLCQVIQMELDHERMSYLLYQMLCGIKHLHSAGIIHRDLKPSNIVVKSDCTLKILDFGLARTAGTSFMMTPYVVTRYYRAPEVILGMGYKENVDIWSVGCIMGEMVRHKILFPGRDYIDQWNKVIEQLGTPCPEFMKKLQPTVRNYVENRPKYAGLTFPKLFPDSLFPADSEHNKLKASQARDLLSKMLVIDPAKRISVDDALQHPYINVWYDPAEVEAPPPQIYDKQLDEREHTIEEWKELIYKEVMNSEEKTKNGVVKGQPSPSGAAVNSSESLPPSSSVNDISSMSTDQTLASDTDSSLEASAGPLGCCR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Protein Kinase, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5602
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.