missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JNK2 Recombinant Rabbit Monoclonal Antibody (004)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA541053
This item is not returnable.
View return policy
Description
JNK2 Recombinant Monoclonal Antibody for Western Blot, IP, ELISA
MAPK9 (JNK2) is a serine/threonine kinase which plays an important role in the MAPK pathway. JNK2 (p54a, SAPK1a), along with JNK1 and JNK3, is thought to play an important role in nuclear signal transduction through its environmental stress activation and subsequent phosphorylation of the nuclear transcription factor p53.
Specifications
| JNK2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MAPK9 | |
| AI851083; c-Jun kinase 2; c-Jun N-terminal kinase 2; I79_012561; JNK; JNK/SAPK alpha; JNK1; JNK1A2; JNK2; JNK21B1/2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; JNK-46; JNK-55; Jun kinase; MAP kinase 10; MAP kinase 8; MAP kinase 9; MAPK 10; MAPK 9; MAPK10; MAPK8; Mapk9; mitogen activated protein kinase 9; mitogen-activated protein kinase 9; OTTHUMP00000161542; OTTHUMP00000161543; p54a; p54-alpha; p54aSAPK; PRKM8; PRKM9; protein kinase, mitogen-activated 9; SAPK; SAPK1; SAPK1a; SAPK1c; SAPK-alpha; stress activated protein kinase alpha II; stress-activated protein kinase 1a; Stress-activated protein kinase JNK2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5601 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 004 | |
| PBS with no preservative | |
| P45984 | |
| MAPK9 | |
| Recombinant Human JNK2/MAPK9 Protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction