missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JNK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517634
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
MAPK9 (JNK2) is a serine/threonine kinase which plays an important role in the MAPK pathway. JNK2 (p54a, SAPK1a), along with JNK1 and JNK3, is thought to play an important role in nuclear signal transduction through its environmental stress activation and subsequent phosphorylation of the nuclear transcription factor p53.
Specifications
| JNK2 | |
| Polyclonal | |
| Unconjugated | |
| MAPK9 | |
| AI851083; c-Jun kinase 2; c-Jun N-terminal kinase 2; I79_012561; JNK; JNK/SAPK alpha; JNK1; JNK1A2; JNK2; JNK21B1/2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; JNK-46; JNK-55; Jun kinase; MAP kinase 10; MAP kinase 8; MAP kinase 9; MAPK 10; MAPK 9; MAPK10; MAPK8; Mapk9; mitogen activated protein kinase 9; mitogen-activated protein kinase 9; OTTHUMP00000161542; OTTHUMP00000161543; p54a; p54-alpha; p54aSAPK; PRKM8; PRKM9; protein kinase, mitogen-activated 9; SAPK; SAPK1; SAPK1a; SAPK1c; SAPK-alpha; stress activated protein kinase alpha II; stress-activated protein kinase 1a; Stress-activated protein kinase JNK2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100764964, 26420, 50658, 5601 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 9 μg/mL | |
| 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P45984, P49186, Q9WTU6 | |
| MAPK9 | |
| Synthetic peptide corresponding to human JNK2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey, Hamster | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction