missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JNK2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | JNK2 |
|---|---|
| Dilution | ELISA 1:5000-1:10000, Immunoprecipitation 0.5-2 uL/mg of lysate |
| Applications | ELISA, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18668365
|
Novus Biologicals
NBP2-99547-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18628075
|
Novus Biologicals
NBP2-99547-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
JNK2 Polyclonal antibody specifically detects JNK2 in Human samples. It is validated for ELISA, ImmunoprecipitationSpecifications
| JNK2 | |
| ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer, Hypoxia, Immunology, Innate Immunity, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
| 0.2 um filtered solution in PBS | |
| 5601 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000, Immunoprecipitation 0.5-2 uL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| c-Jun kinase 2, c-Jun N-terminal kinase 2, EC 2.7.11, EC 2.7.11.24, JNK2A, JNK2BETA, JNK2JNK2ALPHA, JNK-55, Jun kinase, MAP kinase 9, MAPK 9, mitogen-activated protein kinase 9, p54a, p54aSAPK, PRKM9JNK2B, SAPK, Stress-activated protein kinase JNK2 | |
| Produced in rabbits immunized with purified, recombinant Human JNK2 (Accession#: NP_002743.3; Met1-Arg424) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title