missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JNK1/JNK2 Monoclonal Antibody (252323)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561095
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
C-Jun N-terminal Kinase (JNK) is also known as Mitogen-activated protein kinase (MAPK8). It belongs to the MAPK superfamily of stress-activated protein kinases. MAPKs are Serine-threonine protein kinases that are activated in response to a variety of extracellular stimuli, and mediate signal transduction from the cell surface to the nucleus. JNK is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. JNK pathways are activated by stress and inflammatory signals. JNK is expressed as ten different isoforms due to differential mRNA splicing. The predominant forms are JNK1 and JNK2.
Specifications
| JNK1/JNK2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P45983, P45984, P49185, P49186, Q91Y86, Q9WTU6 | |
| MAPK8, MAPK9 | |
| E. coli-derived recombinant human JNK1 isoform 2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Western Blot, Immunocytochemistry | |
| 252323 | |
| Unconjugated | |
| MAPK9 | |
| AI849689; AI851083; C-jun kinase; c-Jun kinase 2; c-Jun N-terminal kinase 1; c-Jun N-terminal kinase 2; I79_012561; JNK; JNK 55; JNK/SAPK alpha; JNK1; JNK1 beta1 protein kinase; JNK1A2; JNK2; JNK21B1/2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; JNK-46; JNK-55; Jun kinase; JUN N-terminal kinase; MAP kinase 8; MAP kinase 9; MAPK; MAPK 8; MAPK 9; Mapk8; Mapk9; mitogen activated protein kinase 8; mitogen activated protein kinase 9; mitogen-activated protein kinase 8; mitogen-activated protein kinase 8 isoform JNK1 alpha1; mitogen-activated protein kinase 8 isoform JNK1 beta2; mitogen-activated protein kinase 9; OTTHUMP00000019552; p54 gamma; p54a; p54-alpha; p54aSAPK; Phospho-JNK; PRKM; PRKM8; Prkm9; protein kinase JNK1; protein kinase mitogen-activated 8; protein kinase, mitogen-activated 9; SAPK; SAPK 1; SAPK gamma; SAPK1; SAPK1A; SAPK1c; SAPK-alpha; stress activated protein kinase alpha II; stress-activated protein kinase 1; stress-activated protein kinase 1a; Stress-activated protein kinase 1c; stress-activated protein kinase JNK1; Stress-activated protein kinase JNK2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 116554, 26419, 26420, 50658, 5599, 5601 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction