missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JNK1/JNK2/JNK3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538289
This item is not returnable.
View return policy
Description
The human JNK1 (PRKM8, SAPK1, MAPK8) gene maps to chromosome 10q11.22 and shares 83% amino acid identity with JNK2. JNK1 is necessary for normal activation and differentiation of CD4 helper T (TH) cells into TH1 and TH2 effector cells. Capsaicin activates JNK1 and p38 in Ras-transformed human breast epithelial cells. Nitrogen oxides (NOx) upregulate JNK1 in addition to c-Fos, c-Jun and other signaling kinases, including MEKK1 and p38. JNK3 (MK10, MAPK10, PRKM10) is activated by pro-inflammatory cytokines and environmental stresss by phosphorylating transcription factors such as c-Jun and ATF2. This is important for AP-1 transcriptional activity regulation. JNK3 is crucial for neuronal apoptosis (stress-induced).
Specifications
| JNK1/JNK2/JNK3 | |
| Polyclonal | |
| Unconjugated | |
| MAPK9 | |
| JNK 46, JNK 55, JNK, JNK1, JNK1A2, JNK2, JNK21B1/2, JNK2ALPHA, JNK2B, JNK2BETA, JNK3, JNK3A, MAP kinase 8, MAP kinase 9, MAP kinase p49 3F12, MAPK 8, MAPK 9, MAPK10, MAPK8, MAPK9, MGC50974, Mitogen activated protein kinase 10, Mitogen activated protein kinase 8, Mitogen activated protein kinase 9, JNK3; JNK3A; PRKM10; p54bSAPK; MAPK10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116554, 25272, 26414, 26419, 26420, 50658, 5599, 5601, 5602 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P45983, P45984, P53779, Q61831, Q91Y86, Q9WTU6, P49185, P49186, P49187 | |
| MAPK10, MAPK8, MAPK9 | |
| A synthetic non-phosphopeptide derived from human JNK1/2/3 around the phosphorylation site of Thr183/Tyr185 (M-M-TP-P-YP-V-V) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction