missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX JNK1α1/SAPK1c Protein, Unactive

Product Code. p-7228362
Encompass_Preferred
Click to view available options
Quantity:
250 μg
50 μg
Unit Size:
250µg
50µg
This item is not returnable. View return policy

Product Code. 17340134

Brand: Eurofins DiscoverX 14328

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Unactive, N-terminal His6-tagged recombinant, human, full-length JNK1α1/SAPK1c, for use in Kinase Assays.

c-Jun N-terminal kinases (JNKs), a stress-activated protein kinase (SAPK), is involved in cellular responses to environmental stresses. JNK kinases can be activated by stimuli such as UV light, radiation, protein synthesis inhibitors, ceramide, DNA-damaging drugs, chemopreventive drugs, TNF-a, and interleukin 1. JNK binds to an NH2-terminal region of ATF2 and c-Jun and phosphorylates two sites within the activation domain of each transcription factor. Three JNK genes (Jnk1, -2 and -3) have been identified in humans; however, splice variants result in a total of 10 isoforms. Therapeutic inhibition of JNK may provide clinical benefit in disease as diverse as immune inflammatory (joint, bowel, pulmonary), ischemic injury (brain, cardiac) and neurodegenerative (Alzheimer's, Parkinson's) diseases.

EMBL L26318

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 0.1 mM EGTA, 150 mM NaCl, 0.03% Brij-35, 50% glycerol, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptothanol. Liquid at -20°C.
Molecular Weight (g/mol) 45 kDa
Quantity 50 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Full-length
Purity or Quality Grade ≥69%
Protein JNK1a1, unactive
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.