missing translation for 'onlineSavingsMsg'
Learn More

JNK Inhibitor VIII, MedChemExpress

Product Code. 16451089
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16451089

Brand: Medchem Express HY1075981mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress JNK Inhibitor VIII (TCS JNK 6o) is a c-Jun N-terminal kinases (JNK-1, -2, and -3) inhibitor with Ki values of 2 nM, 4 nM, 52 nM, respectively, and has IC50 values of 45 nM and 160 nM for JNK-1 and -2, respectively.

JNK Inhibitor VIII (TCS JNK 6o) is a c-Jun N-terminal kinases (JNK-1, -2, and -3) inhibitor with Ki values of 2 nM, 4 nM, 52 nM, respectively, and has IC50 values of 45 nM and 160 nM for JNK-1 and -2, respectively.

Specifications

Chemical Name or Material JNK Inhibitor VIII
CAS 894804-07-0
Color White
Purity Grade Notes Research
Molecular Formula C18H20N4O4
Quantity 1 mg
Synonym TCS JNK 6o
Solubility Information DMSO : 250 mg/mL (701.50 mM; Need ultrasonic)
SMILES N#CC1=C(N=C(NC(CC2=C(C=CC(OC)=C2)OC)=O)C=C1N)OCC
Molecular Weight (g/mol) 356.38
Formula Weight 356.38
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.