missing translation for 'onlineSavingsMsg'
Learn More

JNK-IN-7, MedChemExpress

Product Code. 16406846
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16406846

Brand: Medchem Express HY15617

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress JNK-IN-7 is a potent JNK inhibitor with IC50 of 1.5, 2 and 0.7 nM for JNK1, JNK2 and JNK3, respectively.

JNK-IN-7 is a potent JNK inhibitor with IC50 of 1.5, 2 and 0.7 nM for JNK1, JNK2 and JNK3, respectively.

Specifications

Chemical Name or Material JNK-IN-7
Physical Form Solid
CAS 1408064-71-0
Color Gray
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C28H27N7O2
For Use With (Application) Cancer-Kinase/protease
Synonym JNK inhibitor
Solubility Information DMSO : 33.33 mg/mL (67.53 mM; Need ultrasonic)
SMILES O=C(C1=CC(NC(/C=C/CN(C)C)=O)=CC=C1)NC(C=C2)=CC=C2NC3=NC=CC(C4=CN=CC=C4)=N3
Molecular Weight (g/mol) 493.56
Formula Weight 493.56
Percent Purity 98.41%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.