missing translation for 'onlineSavingsMsg'
Learn More

JNJ16259685, MedChemExpress

Product Code. 16465737
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16465737

Brand: Medchem Express HY100407

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress JNJ16259685 is a selective antagonist of mGlu1 receptor, and inhibits the synaptic activation of mGlu1 in a concentration-dependent manner with IC50 of 19 nM.

JNJ16259685 is a selective antagonist of mGlu1 receptor, and inhibits the synaptic activation of mGlu1 in a concentration-dependent manner with IC50 of 19 nM.

Specifications

Chemical Name or Material JNJ16259685
Physical Form Solid
CAS 409345-29-5
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H23NO3
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 100 mg/mL (307.31 mM)
SMILES O=C(C1=CC=C2N=C3C(CCCO3)=CC2=C1)[C@H]4CC[C@@H](OC)CC4
Molecular Weight (g/mol) 325.4
Formula Weight 325.4
Percent Purity 98.1%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.