missing translation for 'onlineSavingsMsg'
Learn More

JNJ-7777120, MedChemExpress

Product Code. 16481877
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481877 50 mg 50mg
16478407 5 mg 5mg
16466206 100 mg 100mg
16455718 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16481877 Supplier Medchem Express Supplier No. HY13508
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress JNJ-7777120 is a selective H4R antagonist with Ki of 4 ±1 nM, exhibits >1000-fold selectivity over the other histamin receptors.

JNJ-7777120 is a selective H4R antagonist with Ki of 4 ±1 nM, exhibits >1000-fold selectivity over the other histamin receptors.

Specifications

Chemical Name or Material JNJ-7777120
CAS 459168-41-3
Color White
Purity Grade Notes Research
Molecular Formula C14H16ClN3O
Quantity 50 mg
Solubility Information DMSO : ≥ 50 mg/mL (180.02 mM)
SMILES O=C(C(N1)=CC2=C1C=CC(Cl)=C2)N3CCN(C)CC3
Molecular Weight (g/mol) 277.75
Formula Weight 277.75
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.