missing translation for 'onlineSavingsMsg'
Learn More

JNJ-64619178, MedChemExpress

Product Code. 16430779
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16430779

Brand: Medchem Express HY101564

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress JNJ-64619178 (Onametostat) is a selective, orally active and pseudo-irreversible protein arginine methyltransferase 5 (PRMT5) inhibitor with an IC50 of 0.14 nM. JNJ-64619178 has potent activity In lung cancer.

JNJ-64619178 (Onametostat) is a selective, orally active and pseudo-irreversible protein arginine methyltransferase 5 (PRMT5) inhibitor with an IC50 of 0.14 nM. JNJ-64619178 has potent activity In lung cancer.

Specifications

Chemical Name or Material JNJ-64619178
CAS 2086772-26-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H23BrN6O2
Quantity 1 mg
Synonym Onametostat
Solubility Information DMSO : 125 mg/mL (258.61 mM; Need ultrasonic)
SMILES BrC1=C(N)N=C(C=C(CC[C@@H]2[C@@H](O)[C@@H](O)[C@H](N3C(N=CN=C4N)=C4C=C3)C2)C=C5)C5=C1
Molecular Weight (g/mol) 483.36
Formula Weight 483.36
Percent Purity 98.56%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.