missing translation for 'onlineSavingsMsg'
Learn More

JNJ-63533054, MedChemExpress

Product Code. 16497736
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16497736

Brand: Medchem Express HY19838

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress JNJ-63533054 is a potent, selective and orally active GPR139 agonist with an EC50 of 16 nM for human GPR139 (hGPR139). JNJ-63533054 shows selective for GPR139 over other GPCRs, ion channels, and transporters. JNJ-63533054 can cross the blood-brain barrier (BBB).

JNJ-63533054 is a potent, selective and orally active GPR139 agonist with an EC50 of 16 nM for human GPR139 (hGPR139). JNJ-63533054 shows selective for GPR139 over other GPCRs, ion channels, and transporters. JNJ-63533054 can cross the blood-brain barrier (BBB).

Specifications

Chemical Name or Material JNJ-63533054
Physical Form Solid
CAS 1802326-66-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C17H17ClN2O2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 50 mg/mL (157.84 mM)
SMILES O=C(NCC(N[C@H](C1=CC=CC=C1)C)=O)C2=CC=CC(Cl)=C2
Molecular Weight (g/mol) 316.78
Formula Weight 316.78
Percent Purity 99.38%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.