Learn More
ACROBiosystems Biotinylated Human Integrin alpha 4 beta 7 (ITGA4&ITGB7) Heterodimer Protein, His, Avitag™&Tag Free
Description
Integrin alpha 4 beta 7 is an integrin family adhesion receptor that shares subunits with alpha 4 beta 1 (VLA4) and the E-Cadherin receptor, alpha E beta 7 . Integrin alpha 4 beta 7, which is critical for homing to intestinal mucosa, is induced during T cell activation in Peyer’s patches or mesenteric lymph nodes.Its expression requires signals from local dendritic and stromal cells, including secreted retinoic acid. The HIV-1 envelope protein gp120 binds to the active form of Integrin alpha 4 beta 7, and this may or may not account for the concentration of HIV-1 virus in the gut-associated lymphoid tissue (GALT). Integrin alpha 4 beta 7 may also be involved in lymphocyte trafficking in acute intestinal graft vs. host disease (GVHD).
Specifications
Specifications
| Accession Number | NP 000876.3 (ITGA4) and NP_000880.1 (ITGB7) |
| Molecular Weight (g/mol) | 113.2 kDa (ITGA4) and 81.8 kDa (ITGB7) |
| Quantity | 200 μg |
| Source | Biotinylated Human ITGA4&ITGB7; Heterodimer Protein, His,Avitag&Tag; Free (IT7-H82W9) is expressed from human 293 cells (HEK293). It contains AA Tyr 34 - Thr 977 (ITGA4) and Glu 20 - His 723 (ITGB7) (Accession # P13612-1 (ITGA4) and P26010-1 (ITGB7)). |
| Immunogen | Tyr 34 - Thr 977 (ITGA4) and Glu 20 - His 723 (ITGB7) |
| Storage Requirements | -20°C |
| Gene Alias | Integrin alpha 4 beta 7,ITGA4andITGB7 |
| Common Name | Integrin alpha 4 beta 7 |
| Conjugate | Precision Avi |
| Species | Human |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.