missing translation for 'onlineSavingsMsg'
Learn More

JNJ-39758979, MedChemExpress

Product Code. 16466017
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466017 5 mg 5mg
16454546 100 mg 100mg
16440739 1 mg 1mg
16433298 10 mg 10mg
16429836 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16466017 Supplier Medchem Express Supplier No. HY101189
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress JNJ-39758979 is a selective, orally active, and high-affinity histamine H4 receptor antagonist with Kis of 12.5, 5.3, and 25 nM for human, mouse, and monkey histamine H4 receptor, respectively. JNJ-39758979 functionally antagonizes histamine-induced cAMP inhibition with a pA2 of 7.9 in transfected cells. JNJ-39758979 shows good anti-inflammatory and antipruritic activity.

JNJ-39758979 is a selective, orally active, and high-affinity histamine H4 receptor antagonist with Kis of 12.5, 5.3, and 25 nM for human, mouse, and monkey histamine H4 receptor, respectively. JNJ-39758979 functionally antagonizes histamine-induced cAMP inhibition with a pA2 of 7.9 in transfected cells. JNJ-39758979 shows good anti-inflammatory and antipruritic activity.

Specifications

Chemical Name or Material JNJ-39758979
Physical Form Solid
CAS 1046447-90-8
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C11H19N5
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 33.33 mg/mL (150.61 mM; Need ultrasonic)
SMILES NC1=NC(C(C)C)=CC(N2C[C@H](N)CC2)=N1
Molecular Weight (g/mol) 221.3
Formula Weight 221.3
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.