missing translation for 'onlineSavingsMsg'
Learn More

JNJ-31020028, MedChemExpress

Product Code. 16458677
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16458677 5 mg 5mg
16426078 10 mg 10mg
16412217 50 mg 50mg
16406476 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16458677 Supplier Medchem Express Supplier No. HY14450
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress JNJ-31020028 is a selective brain penetrant antagonist of neuropeptide Y2 receptor with high affinity (pIC50=8.07, human; pIC50=8.22 rat); >100-fold selective versus human Y1/Y4/Y5 receptors.

JNJ-31020028 is a selective brain penetrant antagonist of neuropeptide Y2 receptor with high affinity (pIC50=8.07, human

Specifications

Chemical Name or Material JNJ-31020028
CAS 1094873-14-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C34H36FN5O2
Quantity 5 mg
Solubility Information DMSO : 21.5 mg/mL (38.01 mM; Need ultrasonic and warming)
SMILES O=C(N(CC)CC)C(N1CCN(C2=CC=C(NC(C3=C(C4=CC=CN=C4)C=CC=C3)=O)C=C2F)CC1)C5=CC=CC=C5
Molecular Weight (g/mol) 565.68
Formula Weight 565.68
Percent Purity 96.07%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.