missing translation for 'onlineSavingsMsg'
Learn More

JNJ 1661010, Tocris Bioscience™

Product Code. 18074312
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18074312 10 mg 10mg
18005102 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18074312 Supplier Tocris Bioscience Supplier No. 3262/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective, reversible FAAH inhibitor

Selective, reversible inhibitor of fatty acid amide hydrolase (FAAH) (IC50 = 12 nM). Brain penetrant and active in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 681136-29-8
Molecular Formula C19H19N5OS
Molecular Weight (g/mol) 365.46
MDL Number MFCD00209157
InChI Key BHBOSTKQCZEAJM-UHFFFAOYSA-N
Synonym n-phenyl-4-3-phenyl-1,2,4-thiadiazol-5-yl piperazine-1-carboxamide, 1-piperazinecarboxamide, n-phenyl-4-3-phenyl-1,2,4-thiadiazol-5-yl, n-phenyl-4-3-phenyl-1,2,4-thiadiazol-5-yl-1-piperazinecarboxamide, acmc-20emff, d0w5wg, jnj hplc, n1-phenyl-4-3-phenyl-1,2,4-thiadiazol-5-yl piperazine-1-carboxamide
PubChem CID 2809273
IUPAC Name N-phenyl-4-(3-phenyl-1,2,4-thiadiazol-5-yl)piperazine-1-carboxamide
SMILES O=C(NC1=CC=CC=C1)N1CCN(CC1)C1=NC(=NS1)C1=CC=CC=C1

Specifications

Quantity 10 mg
Formula Weight 365.45
Percent Purity >99%
Chemical Name or Material JNJ 1661010
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.