missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMY Antibody (HMY117), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50457JF549
This item is not returnable.
View return policy
Description
JMY Monoclonal specifically detects JMY in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| JMY | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| JMY | |
| Synthetic peptide from the c terminus of the peptide sequence, identical for both isoform 1 and 2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| HMY117 | |
| Western Blot, Immunohistochemistry | |
| FLJ37870, junction mediating and regulatory protein, p53 cofactor, junction-mediating and regulatory protein, junction-mediating and -regulatory protein, MGC163496, WAS protein homology region 2 domain containing 1-like 3, WHDC1L3 | |
| Mouse | |
| Protein A purified | |
| Apoptosis | |
| 133746 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction