missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMJD8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£168.00 - £366.00
Specifications
| Antigen | JMJD8 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18383235
|
Novus Biologicals
NBP3-04630-20UL |
20 μL |
£168.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18392017
|
Novus Biologicals
NBP3-04630-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
JMJD8 Polyclonal specifically detects JMJD8 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifica
| JMJD8 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 339123 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| jmjC domain-containing protein 8, C16orf20, jumonji domain containing 8, PP14397 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-285 of human JMJD8 (NP_001005920.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto