missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JMJD8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA520810
This item is not returnable.
View return policy
Description
A suggested positive control is rat kidney tissue lysate. PA5-20810 can be used with blocking peptide PEP-0924.
The jumonji domain-containing protein (JMJD) family is defined by the presence of the JmjC domain that is observed in several diverse species. While several JMJD proteins have been identified as being involved in chromatin regulation, histone demethylation and development, the function of JMJD8 has not been identified.
Specifications
| JMJD8 | |
| Polyclonal | |
| Unconjugated | |
| JMJD8 | |
| 2610003J06Rik; C16orf20; jmjC domain-containing protein 8; Jmjd8; jumonji domain containing 8; jumonji domain-containing protein 8; PP14397; RGD1307381 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 339123, 360498, 72106 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q3TA59, Q6AY40, Q96S16 | |
| JMJD8 | |
| An 18 amino acid peptide from near the carboxy terminus of human JMJD8. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction