missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMJD8 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-77076CL1
This item is not returnable.
View return policy
Description
JMJD8 Polyclonal antibody specifically detects JMJD8 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| JMJD8 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 339123 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| jmjC domain-containing protein 8, C16orf20, jumonji domain containing 8, PP14397 | |
| Antibody was raised against an 18 amino acid synthetic peptide from near the carboxy terminus of human JMJD8. The immunogen is located within amino acids 250 - 300 of JMJD8. Amino Acid Squence: FLYPPEKTPEFHPNKTTL | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction